Cerium(IV) trifluoromethanesulphonate ≥98%
- Environmentally Preferable:
Frequently Bought Together
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Copper(II) trifluoromethanesulphonate ≥99%
: Thermo Fisher Scientific