
Lanthanum(III) trifluoromethanesulphonate, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52093-26-2
:
Formula:C₃F₉LaO₉S₃
MDL Number:MFCD00077560
:586.12 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Related Information
Frequently Bought Together