cis-2,6-Dimethylpiperazine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21655-48-1
:
Formula:C₆H₁₄N₂
MDL Number:MFCD07772435
:114.19 g/mol
Boiling Pt:162 °C
Melting Pt:108…111 °C
Flash Pt:45 °C (113 °F)
UN:1325
ADR:4.1,III
Related Information
Frequently Bought Together