Order Entry
Northern Ireland
ContactUsLinkComponent
Methyl-5-methyl-2-furoate ≥97%
Methyl-5-methyl-2-furoate ≥97%
Catalog # H27570.03
CAS Number:  
Methyl-5-methyl-2-furoate ≥97%
Catalog # H27570.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:2527-96-0
 :
Formula:C₇H₈O₃
MDL Number:MFCD04116374
 :140.14 g/mol
Boiling Pt:255 °C
Density:1,114
Flash Pt:87 °C (189 °F)
Storage Temperature:Ambient