ContactUsLinkComponent
3-Methylcrotonic acid ≥98%
3-Methylcrotonic acid ≥98%
  A16865.14
 :  
3-Methylcrotonic acid ≥98%
  A16865.14
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
 :
 :100.12 g/mol
 :198…200 °C
 :66…68 °C
 :0,969
 :93 °C (199 °F)
 :Ambient
 :208-782-7
 :3261
 :8,III
 :GHS05
 :541-47-9
 :C₅H₈O₂
 :MFCD00004366