
Bis(pentafluorophenyl)phenylphosphine ≥97%
Catalog # A11014.01
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:442.2 g/mol
Boiling Pt:105 °C (0,3 mmHg)
Melting Pt:59…64 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:225-780-1
:GHS07
CAS Number:5074-71-5
Formula:C₁₈H₅F₁₀P
MDL Number:MFCD00000291