Zinc (II) cyclohexanebutyrate dihydrate for atomic absorption spectroscopy
Catalog # 39734.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:77…80 °C
:439.91 g/mol
Storage Temperature:Ambient
EINECS:254-017-5
CAS Number:38582-18-2
Formula:C₂₀H₃₄O₄Zn·2H₂O
MDL Number:MFCD00035537