
Copper(II) trifluoromethanesulphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34946-82-2
:
Formula:C₂CuF₆O₆S₂
MDL Number:MFCD00077492
:361.69 g/mol
Storage Temperature:Ambient
EINECS:252-300-8
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to light grey to light blue Powder
- Titration with Na2S2O3:97.5 to 102.5 %
- HPLC:≥97.5 %
- Titration with TBAH:≥97.5 %
Specification Test Results
Frequently Bought Together