

2-Picolyl bromide hydrobromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31106-82-8
:
Formula:C₆H₇Br₂N
MDL Number:MFCD01863544
:252.94 g/mol
Melting Pt:149…152 °C
UN:2923
ADR:8,II
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to pink to orange-brown Powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Titration with HClO4:97.5 to 102.5 %
Specification Test Results
Frequently Bought Together