Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) for biochemistry
 
undefined
 
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) for biochemistry

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1 % (K.F.)
  • UV:
    (0.1 M in water)
  • pH:
    4.3 to 5.3 (1 % in water at 25°C)
  • Titration with NaOH:
    ≥99 %

 

 

Frequently Bought Together