(±)-α-Terpineol 95% (α-isomer) (mixture of alpha- and gamma-terpineol)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-55-5
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00001557
:154.25 g/mol
Melting Pt:18 °C
Flash Pt:90 °C
EINECS:232-268-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4800 to 1.4850 (20°C, 589 nm)
- Density:(20°C) 0.930 to 0.938 g/ml
- GC:≥94 % (alpha + gamma)
Specification Test Results
Frequently Bought Together