




Diethylaluminium chloride 1 M in hexanes, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-10-6
:
Formula:C₄H₁₀AlCl
MDL Number:MFCD00000459
:120.56 g/mol
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:202-477-2
UN:3394
ADR:4.2,I
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
Specification Test Results
- Appearance:Colorless Solution
- Strength:0.95 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together