Order Entry
Northern Ireland
ContactUsLinkComponent
 
2-Furohydrazide 98%
 
undefined
2-Furohydrazide 98%

 

  • Appearance:
    Beige Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 80 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together