Cyclopentanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-41-3
:
Formula:C₅H₁₀O
MDL Number:MFCD00001363
:86.13 g/mol
Merck Index:14,2742
Boiling Pt:139…141 °C
Melting Pt:–19 °C
Density:0,950
Flash Pt:51 °C (123 °F)
Storage Temperature:Ambient
EINECS:202-504-8
UN:2244
ADR:3,III
:GHS02
Related Information
Frequently Bought Together