Cyclododecanol for synthesis, Sigma-Aldrich®
Catalog # 8.02675.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:0.04 g/l (20 °C)
- Melting Point:75 - 78 °C
- Molar Mass:184.32 g/mol
- Boiling Point:272 °C (1013 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:138 °C
- Ignition Point:380 °C
Specification Test Results
About this item
:
:184.32 g/mol
Boiling Pt:272,7 °C (1013 hPa)
Melting Pt:75…78 °C
Storage Temperature:Ambient
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:217-031-2
UN:3077
:GHS09
CAS Number:1724-39-6
Formula:C₁₂H₂₄O
MDL Number:MFCD00003717