Order Entry
Northern Ireland
ContactUsLinkComponent
1-Aminocyclohexanecarboxylic acid 98%
Irritant
1-Aminocyclohexanecarboxylic acid 98%
Catalog # ACRO203950250
CAS Number:  
undefined
Irritant
1-Aminocyclohexanecarboxylic acid 98%
Catalog # ACRO203950250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

About this item

 :
 :143.19 g/mol
Boiling Pt:272 °C (1013 hPa)
Melting Pt:>300 °C
Density:1,229 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:220-411-0
 :GHS07
CAS Number:2756-85-6
Formula:C₇H₁₃NO₂
MDL Number:MFCD00001487