
6-Methylquinaldine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:877-43-0
:
Formula:C₁₁H₁₁N
MDL Number:MFCD00006762
:157.22 g/mol
Boiling Pt:266…267 °C
Melting Pt:56…58 °C
Storage Temperature:Ambient
EINECS:212-891-5
:GHS07
Related Information
Frequently Bought Together