8-Quinolinol for synthesis, Sigma-Aldrich®
: Merck
:148-24-3
:
:C₉H₇NO
:MFCD00006807
:145.16 g/mol
:13,04869
:267 °C (1013 hPa)
:71,2 °C
:1,03 g/cm³ (20 °C)
:2811
:6.1,III
:M-Clarity™ quality level = MQ200
:205-711-1
:GHS06
:GHS05
:GHS08
:GHS09
:Ambient
- Solubility:0.56 g/l (20 °C)
- Melting Point:72.5 - 74.0 °C
- Molar Mass:145.16 g/mol
- Boiling Point:267 °C (1013 hPa)
Frequently Bought Together