Order Entry
Northern Ireland
ContactUsLinkComponent
8-Quinolinol ACS, Reag. Ph. Eur. analytical reagent, Supelco®
 
 
 
 
8-Quinolinol ACS, Reag. Ph. Eur. analytical reagent, Supelco®
  1.07098.0250
 :  Merck
 :  
undefined
 
 
 
 
8-Quinolinol ACS, Reag. Ph. Eur. analytical reagent, Supelco®
  1.07098.0250
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Solubility:
    0.56 g/l (20 °C)
  • Melting Point:
    72.5 - 74.0 °C
  • Molar Mass:
    145.16 g/mol
  • Boiling Point:
    267 °C (1013 hPa)

 

 

 :
 :145.16 g/mol
 :13,04869
 :267 °C (1013 hPa)
 :71,2 °C
 :1,03 g/cm³ (20 °C)
 :Ambient
 :205-711-1
 :2811
 :6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
 :GHS08
 :GHS09
 :148-24-3
 :C₉H₇NO
 :MFCD00006807