
3-Aminoquinoline
Catalog # MOLEM75990279
Supplier: Molekula
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:144.18 g/mol
Boiling Pt:311 °C (1013 hPa)
Melting Pt:84 °C
EINECS:209-455-1
:GHS07
Storage Temperature:Ambient
CAS Number:580-17-6
Formula:C₉H₈N₂
MDL Number:MFCD00006772
Boiling Pt:311 °C (1013 hPa)