Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
m-Ethyltoluene
m-Ethyltoluene ≥98%
m-Ethyltoluene ≥98%
:
Thermo Fisher Scientific
m-Ethyltoluene ≥98%
:
Thermo Fisher Scientific
:
620-14-4
:
:
C₉H₁₂
:
MFCD00009259
:
120.19 g/mol
:
158…159 °C
:
–96 °C
:
0,865
:
38 °C (100 °F)
:
Ambient
:
210-626-8
:
3295
:
3,III
:
GHS02
:
GHS09
msds_ALFAL18788
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
: Merck
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
: VWR Chemicals
Tetrachloroethylene ≥99%, GPR RECTAPUR®
: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 40…60 °C, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Methanol, SupraSolv® for gas chromatography, Supelco®
: Merck
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
BIODIESEL BLANK -500ML 1 * 1 items
: PerkinElmer