Container, wide neck
Supplier: BURKLE
Certificates
About this item
HDPE, translucent.
- Can be closed tightly
- Good chemical resistance
- Good temperature resistance
Storage or transport container. For liquids, powders or granules.
Caution: Keep the container upright when transporting liquids.
Frequently Bought Together