Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
 
m-Xylene for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
m-Xylene for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    0.2 g/l (20 °C)
  • Melting Point:
    -48 °C
  • Molar Mass:
    106.16 g/mol
  • Boiling Point:
    139 °C (1013 hPa)
  • Vapor Pressure:
    8 hPa (20 °C)
  • Flash Point:
    25 °C
  • Explosion Limit:
    1.1 - 7 %(V)
  • Density:
    0.86 g/cm3 (20 °C)
  • Ignition Point:
    525 °C
  • Saturation concentration:
    35 g/m3 (20 °C) Air
  • Evaporation number:
    13.5
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

 

Frequently Bought Together