
3-Nitro-o-xylene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:83-41-0
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007162
:151.16 g/mol
Boiling Pt:244…246 °C
Melting Pt:13…15 °C
Density:1,129
Flash Pt:107 °C (224 °F)
Storage Temperature:Ambient
EINECS:201-474-3
UN:1665
ADR:6.1,II
:GHS07
Related Information
Frequently Bought Together