
3-Fluoro-o-xylene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:443-82-3
:
Formula:C₈H₉F
MDL Number:MFCD00000323
Boiling Pt:147…148 °C
Density:0,992
Flash Pt:36 °C (96 °F)
Storage Temperature:Ambient
EINECS:207-140-3
UN:1993
ADR:3,III
:124.16 g/mol
:GHS02
Related Information
Frequently Bought Together