£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, Naphthalene 100 µg/ml in cyclohexane
Organic reference standard, Naphthalene 100 µg/ml in cyclohexane
Organic reference standard, Naphthalene 100 µg/ml in cyclohexane
PROONE5056
:
LGC Standards PROMOCHEM
:
Organic reference standard, Naphthalene 100 µg/ml in cyclohexane
PROONE5056
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,5 mL
CAS No.:
91-20-3
Description:
Naphthalene 100 µg/ml in cyclohexane
Formula:
C₁₀H₈
Description:
Naphthalene