Order Entry
Northern Ireland
ContactUsLinkComponent
2-Aminobenzenesulphonic acid 95%
Irritant
2-Aminobenzenesulphonic acid 95%
Catalog # H31763.22
CAS Number:  
Irritant
2-Aminobenzenesulphonic acid 95%
Catalog # H31763.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :173.19 g/mol
Melting Pt:>300 °C
EINECS:201-810-9
 :GHS07
Storage Temperature:Ambient
CAS Number:88-21-1
Formula:C₆H₇NO₃S
MDL Number:MFCD00007705