
N-Methylmorpholine N-oxide monohydrate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70187-32-5
:
Formula:C₅H₁₁NO₂·1H₂O
MDL Number:MFCD00149388
:135.16 g/mol
Melting Pt:73…76 °C
EINECS:231-391-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together