ContactUsLinkComponent
Tetramethylammonium hydrogen sulphate 99+% for HPLC
Irritant
Tetramethylammonium hydrogen sulphate 99+% for HPLC
Certificates
undefined
Irritant
Tetramethylammonium hydrogen sulphate 99+% for HPLC
Certificates
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1 % (K.F.)
  • UV:
    at 200 nm A: ≤0.15
  • Titration with NaOH:
    ≥99.0 % (On dry substance)