4-(Dimethylamino)benzonitrile 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1197-19-9
:
Formula:C₉H₁₀N₂
MDL Number:MFCD00001815
Melting Pt:70…76 °C
EINECS:214-819-8
ADR:6.1,III
:146.19 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:70 to 76 °C
- Titration with HClO4:≥94 %
- Stabilizer:About 500 ppm 4-tert-Butylcatecholo
Specification Test Results
Frequently Bought Together