3-Methylbiphenyl 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:643-93-6
:
Formula:C₁₃H₁₂
MDL Number:MFCD00008533
:168.24 g/mol
Boiling Pt:272 °C
Melting Pt:4…5 °C
Density:1,014
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:211-404-3
Related Information
Frequently Bought Together