4-Phenylbutyrophenone 98%
Catalog # ACRO232820010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:55 to 59 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:224.3 g/mol
Boiling Pt:155 °C (0,2 mmHg)
Melting Pt:55…59 °C
EINECS:226-471-4
CAS Number:5407-91-0
Formula:(C₆H₅)C(O)CH₂CH₂CH₂(C₆H₅)
MDL Number:MFCD00022005