1,1-Diphenylacetone
Catalog # B20494.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:210.28 g/mol
Boiling Pt:111…126 °C (0,1 mmHg)
Melting Pt:59…62 °C
Storage Temperature:Refrigerator
EINECS:212-307-9
CAS Number:781-35-1
Formula:(C₆H₅)₂CHC(O)CH₃
MDL Number:MFCD00008759