Order Entry
Northern Ireland
ContactUsLinkComponent
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Assay (on anhydrous substance):
    Min. 99.90 %
  • Carbonisable substances:
    Passes test
  • IR Spectrum:
    Passes test
  • Substances coloured by H2SO4:
    Passes test ACS/ISO
  • Acidity or alkalinity:
    Max. 0.0001 meq/g
  • Boiling point:
    109 - 112 °C
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    0.865 - 0.868
  • Density (20/20):
    0.865 - 0.870
  • Refractive index (20°C):
    1.496 - 1.498
  • Benzene:
    Max. 50 ppm
  • Evaporation residue:
    Max. 10 ppm
  • Thiophene:
    Max. 1 ppm
  • Total S (as SO4):
    Max. 10 ppm
  • Water:
    Max. 0.03 %
  • Al (Aluminium):
    Max. 0.05 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.02 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.01 ppm
  • Cu (Copper):
    Max. 0.01 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.05 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Ni (Nickel):
    Max. 0.01 ppm
  • Pb (Lead):
    Max. 0.01 ppm
  • Sn (Tin):
    Max. 0.05 ppm
  • Sr (Strontium):
    Max. 0.02 ppm
  • Zn (Zinc):
    Max. 0.01 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together