Dodecylbenzene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-01-3
:
Formula:C₁₈H₃₀
MDL Number:MFCD00008974
Boiling Pt:288…300 °C (760 mmHg)
Melting Pt:–7 °C
Density:0,85 g/cm³
Flash Pt:124 °C
:246.44 g/mol
Storage Temperature:Ambient
EINECS:204-591-8
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4810 to 1.4830 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together