
2-Fluoro-5-(trifluoromethyl)anisole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:261951-78-4
:
Formula:C₈H₆F₄O
MDL Number:MFCD01631564
:194.13 g/mol
Boiling Pt:151 °C (1013 hPa)
Melting Pt:–15 °C
Flash Pt:62 °C (144 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together