3-Chloro-2-methylanisole ≥97%
:3260-88-6
:
:C₈H₉ClO
:MFCD00070772
:156.61 g/mol
:212…215 °C
:–3 °C
:1,160
:92 °C (197 °F)
:Ambient
:221-862-6
:3,III
:GHS02
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.