2-Nitrobenzyl chloride for synthesis, Sigma-Aldrich®
8.41296.0010
: Merck
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Solubility:(20 °C) insoluble
- Melting Point:46 - 49 °C
- Molar Mass:171.58 g/mol
- Boiling Point:127 - 133 °C (13 hPa)
- Flash Point:112 °C
:
:171.58 g/mol
:263 °C (1013 hPa)
:49…51 °C
:1,5557 g/cm³ (20 °C)
:Ambient
:210-300-5
:3457
:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
:612-23-7
:C₇H₆ClNO₂
:MFCD00007185