

4-Chloro-3-nitrobenzotrifluoride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-17-5
:
Formula:C₇H₃ClF₃NO₂
MDL Number:MFCD00007084
:225.55 g/mol
Boiling Pt:221…223 °C
Melting Pt:–3 °C
Density:1,535
Flash Pt:101 °C (213 °F)
Storage Temperature:Ambient
EINECS:204-451-6
UN:2307
ADR:6.1,II
:GHS06
:GHS07
Related Information
Frequently Bought Together