Order Entry
Northern Ireland
ContactUsLinkComponent
6-Hydroxy-m-toluic acid 98%
Irritant
6-Hydroxy-m-toluic acid 98%
Certificates
undefined
Irritant
6-Hydroxy-m-toluic acid 98%

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 155 °C
  • Titration with NaOH:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together