4-tert-Butylbenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-73-7
:
Formula:C₁₁H₁₄O₂
MDL Number:MFCD00002563
:178.23 g/mol
Boiling Pt:283 °C (1013 hPa)
Melting Pt:165…167 °C
Density:0,6 g/cm³ (20 °C)
Flash Pt:180 °C
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:202-696-3
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.3 g/l (20 °C)
- Melting Point:165 - 167 °C
- Molar Mass:178.23 g/mol
- Bulk Density:340 kg/m3
- Boiling Point:280 °C (decomposition)
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:180 °C
- Density:0.6 g/cm3 (20 °C)
- pH:3.9 (H2O) (saturated solution)
- Ignition Point:510 °C
Specification Test Results
Frequently Bought Together