Butyrophenone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:495-40-9
:
Formula:C₆H₅C(O)(CH₂)₂CH₃
MDL Number:MFCD00009397
:148.2 g/mol
Boiling Pt:220 °C (1013 hPa)
Density:0,988 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-799-7
Specification Test Results
- Melting Point:11 - 13 °C
- Molar Mass:148.2 g/mol
- Boiling Point:220 - 222 °C (1013 hPa)
- Flash Point:88 °C
- Density:0.988 g/cm3
- Hill Formula:C10H12O
Specification Test Results
Frequently Bought Together