
2-Bromo-6-nitrotoluene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55289-35-5
:
Formula:C₇H₆BrNO₂
MDL Number:MFCD00009792
:216.03 g/mol
Boiling Pt:108…110 °C (3,3 mmHg)
Melting Pt:37…42 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:259-566-4
:GHS07
Related Information
Frequently Bought Together