Order Entry
Northern Ireland
ContactUsLinkComponent
 
α-Bromo-2-nitrotoluene
Catalog # MOLE28390978-25G
Supplier:  Molekula
CAS Number:  
α-Bromo-2-nitrotoluene
Catalog # MOLE28390978-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :216.03 g/mol
Melting Pt:45…47 °C
Storage Temperature:Refrigerator
EINECS:223-558-9
UN:3261
ADR:8,II
 :GHS05
 :GHS07
CAS Number:3958-60-9
Formula:C₇H₆BrNO₂
MDL Number:MFCD00007184