HPLC columns, Accucore™ aQ
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns are compatible with 100% aqueous mobile phases and offer special selectivity for polar analytes. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub 2 µm particles.
- Ideal for highly aqueous mobile phases
- Retention and resolution of polar analytes
- Polar endcapped C18 stationary phase for alternative selectivity
Limited Time Sale
Select items below are currently on sale
Uniguard™ direct connection guard cartridge holders
Product Information
Frequently Bought Together