1-Chloro-5-(triethylsilyl)-4-pentyne ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:174125-30-5
:
Formula:C₁₁H₂₁ClSi
MDL Number:MFCD00671352
Molecular Weight:216.83 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together