Creatinine, anhydrous 98.5-102%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:60-27-5
:
Formula:C₄H₇N₃O
MDL Number:MFCD00059730
:113.12 g/mol
Melting Pt:295 °C (lit.)
Density:1,09 g/cm³ (lit.)
EINECS:200-466-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:98.5-102.0%
- FTIR:Conforms to Standard
- pH (5% aq soln):7.5-9.0
- Loss on Drying:≤3.0%
- Residue on Ignition:≤0.2%
- Heavy Metals:≤0.001%
Specification Test Results
Frequently Bought Together