2-(4-Hydroxyphenylazo)benzoic acid
Catalog # PIER28010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:242.23 g/mol
EINECS:216-655-2
CAS Number:1634-82-8
Formula:C₁₃H₁₀N₂O₃
MDL Number:MFCD00002428