
Diglycolic anhydride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4480-83-5
:
Formula:C₄H₄O₄
MDL Number:MFCD00006677
:116.07 g/mol
Melting Pt:90…95 °C
EINECS:224-761-5
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to cream Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:90 to 95 °C
- Titration excess NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together