
Diglycolic anhydride, Sigma-Aldrich®
Catalog # 8.41741.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) soluble,Hydrolysis
- Melting Point:91 - 93 °C
- Molar Mass:116.07 g/mol
- Boiling Point:240 - 241 °C (1013 hPa)
Specification Test Results
About this item
:
:116.07 g/mol
:M-Clarity™ quality level = MQ200
EINECS:224-761-5
:GHS07
Storage Temperature:Ambient
CAS Number:4480-83-5
Formula:C₄H₄O₄
MDL Number:MFCD00006677